siRNA Details
| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033688.1 | NP_001028860.1 | 15 | 2669 | ||
| XM_008771935.1 | XP_008770157.1 | 15 | 2733 | |||
| GenBank mRNA | BC101864.1 | - | 15 | 2669 | 7939139 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsc2| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033688.1 | NP_001028860.1 | 15 | 2669 | ||
| XM_008771935.1 | XP_008770157.1 | 15 | 2733 | |||
| GenBank mRNA | BC101864.1 | - | 15 | 2669 | 7939139 | |