siRNA Details
| Gene Symbol: Kctd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100516.1 | NP_001093986.1 | 4 | 2194 | ||
| GenBank mRNA | CK473534.1 | - | 3 | 294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd1| Gene Symbol: Kctd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100516.1 | NP_001093986.1 | 4 | 2194 | ||
| GenBank mRNA | CK473534.1 | - | 3 | 294 | ||