siRNA Details
| Gene Symbol: RGD1311805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009638.1 | NP_001009638.1 | 2, 3 | 265 | ||
| GenBank mRNA | BC087672.1 | - | 2, 3 | 265 | 7317049 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311805| Gene Symbol: RGD1311805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009638.1 | NP_001009638.1 | 2, 3 | 265 | ||
| GenBank mRNA | BC087672.1 | - | 2, 3 | 265 | 7317049 | |