siRNA Details
| Gene Symbol: Syn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034020.1 | NP_001029192.1 | 5 | 831 | ||
| NM_019159.1 | NP_062032.1 | 5 | 831 | |||
| GenBank mRNA | M27925.1 | - | 5 | 831 | ||
| M27926.1 | - | 5 | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syn2| Gene Symbol: Syn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034020.1 | NP_001029192.1 | 5 | 831 | ||
| NM_019159.1 | NP_062032.1 | 5 | 831 | |||
| GenBank mRNA | M27925.1 | - | 5 | 831 | ||
| M27926.1 | - | 5 | 831 | |||