siRNA Details
| Gene Symbol: Cdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168631.1 | NP_001162102.1 | 12 | 2281 | ||
| XM_006232079.2 | XP_006232141.1 | 12 | 2357 | |||
| XM_006232080.2 | XP_006232142.1 | 12 | 2383 | |||
| GenBank mRNA | AF177676.1 | - | 1 | 31 | ||
| AF468011.1 | - | 3 | 474 | |||

Cdh10
Genomic Map