siRNA Details
| Gene Symbol: Cdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168631.1 | NP_001162102.1 | 2 | 353 | ||
| XM_006232079.2 | XP_006232141.1 | 2 | 429 | |||
| XM_006232080.2 | XP_006232142.1 | 2 | 461 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh10| Gene Symbol: Cdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168631.1 | NP_001162102.1 | 2 | 353 | ||
| XM_006232079.2 | XP_006232141.1 | 2 | 429 | |||
| XM_006232080.2 | XP_006232142.1 | 2 | 461 | |||