siRNA Details
| Gene Symbol: Slc38a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003705.2 | NP_001003705.1 | 8 | 1313 | ||
| XM_006255067.2 | XP_006255129.1 | 10 | 1460 | |||
| GenBank mRNA | AY294328.1 | - | 9 | 1408 | ||
| BC086369.1 | - | 8 | 1313 | 7109456 | ||

Slc38a7
Genomic Map