siRNA Details
| Gene Symbol: Katnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024746.1 | NP_001019917.1 | 8 | 667 | ||
| XM_006255078.1 | XP_006255140.1 | 9 | 1043 | |||
| XM_008772277.1 | XP_008770499.1 | 9 | 984 | |||
| XM_008772278.1 | XP_008770500.1 | 9 | 1115 | |||
| XM_008772279.1 | XP_008770501.1 | 9 | 1166 | |||
| XM_008772280.1 | XP_008770502.1 | 9 | 1044 | |||
| GenBank mRNA | AY953248.1 | - | 8 | 667 | ||
| BC161823.1 | - | 8 | 913 | |||

Katnb1
Genomic Map