siRNA Details
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017124.1 | NP_058820.1 | 6 | 691 | ||
| XM_006228981.2 | XP_006229043.1 | 7 | 884 | |||
| XM_008759326.1 | XP_008757548.1 | 6 | 730 | |||
| GenBank mRNA | BC059144.1 | - | 6 | 732 | 6887407 | |
| FQ230385.1 | - | 6 | 709 | |||
| X53517.1 | - | 6 | 691 | |||

Cd37
Genomic Map