siRNA Details
| Gene Symbol: Cd69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134327.1 | NP_599154.1 | 3, 4 | 455 | ||
| GenBank mRNA | AF440759.1 | - | 1, 2 | 190 | ||
| BC088219.1 | - | 3, 4 | 455 | 7384461 | ||
| GU357488.1 | - | 3, 4 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd69| Gene Symbol: Cd69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134327.1 | NP_599154.1 | 3, 4 | 455 | ||
| GenBank mRNA | AF440759.1 | - | 1, 2 | 190 | ||
| BC088219.1 | - | 3, 4 | 455 | 7384461 | ||
| GU357488.1 | - | 3, 4 | 391 | |||