siRNA Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011926.1 | NP_001011926.1 | 2, 3 | 202 | ||
| XM_006255181.2 | XP_006255243.1 | 2, 3 | 214 | |||
| XM_006255182.2 | XP_006255244.1 | 2, 3 | 192 | |||
| XM_006255183.2 | XP_006255245.1 | 1, 2 | 115 | |||
| XM_006255185.2 | XP_006255247.1 | 2, 3 | 148 | |||
| GenBank mRNA | BC089985.1 | - | 2, 3 | 202 | 7318380 | |

Aktip
Genomic Map