siRNA Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011926.1 | NP_001011926.1 | 5 | 528 | ||
| XM_006255181.2 | XP_006255243.1 | 5 | 540 | |||
| XM_006255182.2 | XP_006255244.1 | 5 | 518 | |||
| XM_006255183.2 | XP_006255245.1 | 4 | 441 | |||
| XM_006255185.2 | XP_006255247.1 | 5 | 474 | |||
| GenBank mRNA | BC089985.1 | - | 5 | 528 | 7318380 | |

Aktip
Genomic Map