siRNA Details
| Gene Symbol: Abcc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199377.1 | NP_955409.1 | 27, 28 | 3795 | ||
| XM_006255241.2 | XP_006255303.1 | 27, 28 | 3936 | |||
| XM_006255242.2 | XP_006255304.1 | 26, 27 | 3733 | |||
| XM_008772375.1 | XP_008770597.1 | 27, 28 | 3959 | |||
| XM_008772376.1 | XP_008770598.1 | 27, 28 | 3956 | |||
| XM_008772377.1 | XP_008770599.1 | 26, 27 | 3870 | |||
| XM_008772378.1 | XP_008770600.1 | 27, 28 | 3950 | |||
| GenBank mRNA | AY188179.1 | - | 27, 28 | 3795 | ||
| AY188180.1 | - | 26, 27 | 3558 | |||

Abcc12
Genomic Map