siRNA Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255413.2 | XP_006255475.1 | 6 | 2585 | ||
| XM_008772544.1 | XP_008770766.1 | 6 | 2585 | |||
| XM_008772545.1 | XP_008770767.1 | 6 | 2585 | |||
| XM_008772546.1 | XP_008770768.1 | 6 | 2585 | |||
| XM_008772547.1 | XP_008770769.1 | 6 | 2585 | |||

Zfp827
Genomic Map