siRNA Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030028.1 | NP_001025199.1 | 13 | 1350 | ||
| XM_008772479.1 | XP_008770701.1 | 12 | 1382 | |||
| XM_008772480.1 | XP_008770702.1 | 12 | 1454 | |||
| XM_008772481.1 | XP_008770703.1 | 13 | 1301 | |||
| XM_008772482.1 | XP_008770704.1 | 12 | 1382 | |||
| GenBank mRNA | BC099761.1 | - | 13 | 1350 | 7930411 | |

Elmo3
Genomic Map