siRNA Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030028.1 | NP_001025199.1 | 19, 20 | 2081 | ||
| XM_008772479.1 | XP_008770701.1 | 18, 19 | 2113 | |||
| XM_008772480.1 | XP_008770702.1 | 18, 19 | 2185 | |||
| XM_008772481.1 | XP_008770703.1 | 19, 20 | 2032 | |||
| GenBank mRNA | BC099761.1 | - | 19, 20 | 2081 | 7930411 | |

Elmo3
Genomic Map