siRNA Details
| Gene Symbol: Fhod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191600.1 | NP_001178529.1 | 21, 22 | 3213 | ||
| XM_008772483.1 | XP_008770705.1 | 22, 23 | 4691 | |||
| XM_008772484.1 | XP_008770706.1 | 20, 21 | 4613 | |||

Fhod1
Genomic Map