siRNA Details
| Gene Symbol: Fhod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191600.1 | NP_001178529.1 | 16 | 2309 | ||
| XM_008772483.1 | XP_008770705.1 | 17 | 3787 | |||
| XM_008772484.1 | XP_008770706.1 | 15 | 3709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fhod1| Gene Symbol: Fhod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191600.1 | NP_001178529.1 | 16 | 2309 | ||
| XM_008772483.1 | XP_008770705.1 | 17 | 3787 | |||
| XM_008772484.1 | XP_008770706.1 | 15 | 3709 | |||