siRNA Details
| Gene Symbol: Atp6v0d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011927.1 | NP_001011927.1 | 6 | 740 | ||
| GenBank mRNA | BC088462.1 | - | 6 | 740 | 7127604 | |
| FQ213716.1 | - | 6 | 756 | |||
| FQ215412.1 | - | 5 | 475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v0d1| Gene Symbol: Atp6v0d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011927.1 | NP_001011927.1 | 6 | 740 | ||
| GenBank mRNA | BC088462.1 | - | 6 | 740 | 7127604 | |
| FQ213716.1 | - | 6 | 756 | |||
| FQ215412.1 | - | 5 | 475 | |||