siRNA Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100660.1 | NP_001094130.1 | 13 | 1521 | ||
| XM_006255470.2 | XP_006255532.1 | 13 | 1870 | |||
| XM_006255471.2 | XP_006255533.1 | 13 | 1982 | |||
| XM_006255472.2 | XP_006255534.1 | 13 | 1971 | |||
| XM_006255473.2 | XP_006255535.1 | 13 | 1871 | |||
| XM_006255474.2 | XP_006255536.1 | 13 | 1870 | |||
| XM_006255475.2 | XP_006255537.1 | 13 | 1534 | |||
| GenBank mRNA | AY724537.1 | - | 5 | 750 | ||
| BC099098.1 | - | 3 | 488 | |||

Fam65a
Genomic Map