siRNA Details
Gene Symbol: Enkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106180.1 | NP_001099650.1 | 7 | 1263 | ||
XM_006255476.2 | XP_006255538.1 | 8 | 1043 | |||
XM_006255477.2 | XP_006255539.1 | 8 | 1093 | |||
XM_006255478.2 | XP_006255540.1 | 8 | 1027 |
