siRNA Details
| Gene Symbol: Tcf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751893.3 | XP_003751941.1 | 1 | 382 | ||
| XM_006255791.2 | XP_006255853.1 | 1 | 382 | |||
| XM_006255792.2 | XP_006255854.1 | 1 | 382 | |||
| XM_006255793.2 | XP_006255855.1 | 1 | 385 | |||
| XM_006255794.1 | XP_006255856.1 | 1 | 382 | |||
| XM_214707.8 | XP_214707.6 | 1 | 382 | |||
| GenBank mRNA | FQ215148.1 | - | 1 | 146 | ||
| FQ230504.1 | - | 1 | 128 | |||

Tcf25
Genomic Map