siRNA Details
| Gene Symbol: Ttc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136162.1 | NP_001129634.1 | 7 | 752 | ||
| XM_006255802.2 | XP_006255864.1 | 9 | 920 | |||
| XM_006255803.2 | XP_006255865.1 | 9 | 920 | |||
| XM_006255805.2 | XP_006255867.1 | 7 | 761 | |||

Ttc13
Genomic Map