siRNA Details
| Gene Symbol: Dclre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106201.1 | NP_001099671.1 | 9 | 3165 | ||
| XM_006231634.1 | XP_006231696.1 | 10 | 3841 | |||
| XM_006231635.2 | XP_006231697.1 | 10 | 3779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dclre1a| Gene Symbol: Dclre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106201.1 | NP_001099671.1 | 9 | 3165 | ||
| XM_006231634.1 | XP_006231696.1 | 10 | 3841 | |||
| XM_006231635.2 | XP_006231697.1 | 10 | 3779 | |||