siRNA Details
| Gene Symbol: Tdrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106202.1 | NP_001099672.1 | 14 | 1983 | ||
| XM_006231642.2 | XP_006231704.1 | 15 | 2246 | |||
| XM_008760524.1 | XP_008758746.1 | 15 | 2053 | |||
| XM_008760525.1 | XP_008758747.1 | 15 | 2018 | |||
| XM_008760526.1 | XP_008758748.1 | 14 | 2183 | |||
| XM_008760528.1 | XP_008758750.1 | 14 | 1812 | |||
| XM_008760529.1 | XP_008758751.1 | 14 | 2090 | |||
| XM_008760530.1 | XP_008758752.1 | 15 | 2246 | |||

Tdrd1
Genomic Map