siRNA Details
| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011929.1 | NP_001011929.1 | 8 | 1247 | ||
| XM_006233431.2 | XP_006233493.1 | 10 | 1373 | |||
| XM_006233433.2 | XP_006233495.1 | 10 | 1323 | |||
| XM_006233435.2 | XP_006233497.1 | 8 | 1259 | |||
| GenBank mRNA | BC079085.1 | - | 8 | 1247 | 7123144 | |
| FQ216102.1 | - | 4 | 461 | |||
| FQ234013.1 | - | 8 | 970 | |||

Sh3glb1
Genomic Map