siRNA Details
Gene Symbol: Znhit6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106203.2 | NP_001099673.1 | 3 | 800 | ||
GenBank mRNA | BC161957.1 | - | 3 | 800 | 9027306 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Znhit6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106203.2 | NP_001099673.1 | 3 | 800 | ||
GenBank mRNA | BC161957.1 | - | 3 | 800 | 9027306 |