siRNA Details
| Gene Symbol: Mcoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039005.1 | NP_001034094.1 | 2 | 184 | ||
| GenBank mRNA | BC099787.1 | - | 2 | 184 | 7934506 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcoln2| Gene Symbol: Mcoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039005.1 | NP_001034094.1 | 2 | 184 | ||
| GenBank mRNA | BC099787.1 | - | 2 | 184 | 7934506 | |