siRNA Details
Gene Symbol: Rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017132.2 | NP_058828.2 | 2 | 417 | ||
GenBank mRNA | BC061962.1 | - | 2 | 371 | 5598390 | |
CB791891.1 | - | 2 | 417 | |||
U15734.1 | - | 2 | 428 |
