siRNA Details
| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106206.1 | NP_001099676.1 | 2 | 501 | ||
| GenBank mRNA | BC158745.1 | - | 2 | 441 | 9100726 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo5| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106206.1 | NP_001099676.1 | 2 | 501 | ||
| GenBank mRNA | BC158745.1 | - | 2 | 441 | 9100726 | |