siRNA Details
Gene Symbol: Fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106206.1 | NP_001099676.1 | 3, 4 | 963 | ||
GenBank mRNA | BC158745.1 | - | 3, 4 | 903 | 9100726 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106206.1 | NP_001099676.1 | 3, 4 | 963 | ||
GenBank mRNA | BC158745.1 | - | 3, 4 | 903 | 9100726 |