siRNA Details
Gene Symbol: RGD1305235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017447.1 | NP_001017447.1 | 1, 2 | 116 | ||
GenBank mRNA | BC078920.1 | - | 1, 2 | 116 | 7096641 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1305235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017447.1 | NP_001017447.1 | 1, 2 | 116 | ||
GenBank mRNA | BC078920.1 | - | 1, 2 | 116 | 7096641 |