siRNA Details
| Gene Symbol: Pabpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106208.2 | NP_001099678.1 | 1 | 713 | ||
| GenBank mRNA | BC168959.1 | - | 1 | 713 | 7135469 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc6| Gene Symbol: Pabpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106208.2 | NP_001099678.1 | 1 | 713 | ||
| GenBank mRNA | BC168959.1 | - | 1 | 713 | 7135469 | |