siRNA Details
| Gene Symbol: T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106209.1 | NP_001099679.1 | 4, 5 | 786 | ||
| XM_006227918.2 | XP_006227980.1 | 6, 7 | 1686 | |||
| XM_008758750.1 | XP_008756972.1 | 3, 4 | 935 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

T| Gene Symbol: T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106209.1 | NP_001099679.1 | 4, 5 | 786 | ||
| XM_006227918.2 | XP_006227980.1 | 6, 7 | 1686 | |||
| XM_008758750.1 | XP_008756972.1 | 3, 4 | 935 | |||