siRNA Details
| Gene Symbol: Ciao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008766.1 | NP_001008766.1 | 4 | 533 | ||
| GenBank mRNA | BC088474.1 | - | 4 | 533 | 7129030 | |
| FQ214368.1 | - | 4 | 556 | |||
| FQ230934.1 | - | 4 | 547 | |||
| FQ231672.1 | - | 4 | 526 | |||

Ciao1
Genomic Map