siRNA Details
| Gene Symbol: Clcn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031568.2 | NP_113756.1 | 15 | 1345 | ||
| GenBank mRNA | Z67744.1 | - | 15 | 1346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcn7| Gene Symbol: Clcn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031568.2 | NP_113756.1 | 15 | 1345 | ||
| GenBank mRNA | Z67744.1 | - | 15 | 1346 | ||