siRNA Details
| Gene Symbol: F10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017143.2 | NP_058839.1 | 8 | 1069 | ||
| GenBank mRNA | BC088151.1 | - | 8 | 1069 | 7383301 | |
| X79807.1 | - | 8 | 981 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F10| Gene Symbol: F10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017143.2 | NP_058839.1 | 8 | 1069 | ||
| GenBank mRNA | BC088151.1 | - | 8 | 1069 | 7383301 | |
| X79807.1 | - | 8 | 981 | |||