siRNA Details
| Gene Symbol: F10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017143.2 | NP_058839.1 | 6 | 783 | ||
| GenBank mRNA | BC088151.1 | - | 6 | 783 | 7383301 | |
| FQ211285.1 | - | 6 | 707 | |||
| FQ219425.1 | - | 6 | 701 | |||
| X79807.1 | - | 6 | 695 | |||

F10
Genomic Map