siRNA Details
Gene Symbol: Gch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024356.1 | NP_077332.1 | 2, 3 | 557 | ||
GenBank mRNA | FQ220953.1 | - | 2, 3 | 544 | ||
M58364.1 | - | 2, 3 | 557 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024356.1 | NP_077332.1 | 2, 3 | 557 | ||
GenBank mRNA | FQ220953.1 | - | 2, 3 | 544 | ||
M58364.1 | - | 2, 3 | 557 |