siRNA Details
| Gene Symbol: Gch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024356.1 | NP_077332.1 | 5, 6 | 718 | ||
| GenBank mRNA | FQ220953.1 | - | 5, 6 | 705 | ||
| M58364.1 | - | 5, 6 | 718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gch1| Gene Symbol: Gch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024356.1 | NP_077332.1 | 5, 6 | 718 | ||
| GenBank mRNA | FQ220953.1 | - | 5, 6 | 705 | ||
| M58364.1 | - | 5, 6 | 718 | |||