siRNA Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033653.1 | NP_387502.1 | 10 | 1074 | ||
| GenBank mRNA | BC166508.1 | - | 10 | 1057 | ||
| FM031063.1 | - | 3 | 158 | |||
| FM132632.1 | - | 6 | 600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maoa| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033653.1 | NP_387502.1 | 10 | 1074 | ||
| GenBank mRNA | BC166508.1 | - | 10 | 1057 | ||
| FM031063.1 | - | 3 | 158 | |||
| FM132632.1 | - | 6 | 600 | |||