siRNA Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033653.1 | NP_387502.1 | 7 | 677 | ||
| GenBank mRNA | BC166508.1 | - | 7 | 660 | ||
| D00688.1 | - | 7 | 683 | |||
| FM117485.1 | - | 6 | 553 | |||
| FM132632.1 | - | 3 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maoa| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033653.1 | NP_387502.1 | 7 | 677 | ||
| GenBank mRNA | BC166508.1 | - | 7 | 660 | ||
| D00688.1 | - | 7 | 683 | |||
| FM117485.1 | - | 6 | 553 | |||
| FM132632.1 | - | 3 | 203 | |||