siRNA Details
| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169142.1 | NP_001162613.1 | 3 | 861 | ||
| XM_008758753.1 | XP_008756975.1 | 3 | 1028 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp12| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169142.1 | NP_001162613.1 | 3 | 861 | ||
| XM_008758753.1 | XP_008756975.1 | 3 | 1028 | |||