siRNA Details
| Gene Symbol: Zfp494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_234948.3 | XP_234948.1 | 3 | 1377 | ||
| Gene Symbol: LOC100362054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758861.1 | XP_008757083.1 | 6 | 2333 | ||

Zfp494
Genomic Map