siRNA Details
| Gene Symbol: Tlr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019178.1 | NP_062051.1 | 3 | 2575 | ||
| GenBank mRNA | AF057025.1 | - | 3 | 2597 | ||
| FQ233814.1 | - | 1 | 629 | |||
| GQ175880.1 | - | 3 | 2575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlr4| Gene Symbol: Tlr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019178.1 | NP_062051.1 | 3 | 2575 | ||
| GenBank mRNA | AF057025.1 | - | 3 | 2597 | ||
| FQ233814.1 | - | 1 | 629 | |||
| GQ175880.1 | - | 3 | 2575 | |||