siRNA Details
| Gene Symbol: Tlr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019178.1 | NP_062051.1 | 1, 2 | 327 | ||
| GenBank mRNA | AF057025.1 | - | 1, 2 | 349 | ||
| EU929065.1 | - | 1, 2 | 52 | |||
| GQ175880.1 | - | 1, 2 | 327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlr4| Gene Symbol: Tlr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019178.1 | NP_062051.1 | 1, 2 | 327 | ||
| GenBank mRNA | AF057025.1 | - | 1, 2 | 349 | ||
| EU929065.1 | - | 1, 2 | 52 | |||
| GQ175880.1 | - | 1, 2 | 327 | |||