siRNA Details
Gene Symbol: Zfp606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228138.2 | XP_006228200.1 | 7 | 842 | ||
XM_006228139.2 | XP_006228201.1 | 6 | 789 | |||
XM_218283.7 | XP_218283.5 | 6 | 806 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228138.2 | XP_006228200.1 | 7 | 842 | ||
XM_006228139.2 | XP_006228201.1 | 6 | 789 | |||
XM_218283.7 | XP_218283.5 | 6 | 806 |