siRNA Details
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228138.2 | XP_006228200.1 | 7 | 955 | ||
| XM_006228139.2 | XP_006228201.1 | 6 | 902 | |||
| XM_218283.7 | XP_218283.5 | 6 | 919 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp606| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228138.2 | XP_006228200.1 | 7 | 955 | ||
| XM_006228139.2 | XP_006228201.1 | 6 | 902 | |||
| XM_218283.7 | XP_218283.5 | 6 | 919 | |||