siRNA Details
| Gene Symbol: Tyms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019179.1 | NP_062052.1 | 7 | 848 | ||
| XM_008767420.1 | XP_008765642.1 | 7 | 1337 | |||
| XM_008767421.1 | XP_008765643.1 | 8 | 893 | |||
| GenBank mRNA | BC126093.1 | - | 7 | 862 | 8362547 | |
| L12138.1 | - | 7 | 848 | |||

Tyms
Genomic Map