siRNA Details
| Gene Symbol: Gltscr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207591.1 | NP_997474.1 | 1 | 18 | ||
| GenBank mRNA | AY535002.1 | - | 1 | 18 | ||
| BC086359.1 | - | 1 | 22 | 7304013 | ||
| FQ212888.1 | - | 1 | 49 | |||
| FQ213201.1 | - | 1 | 28 | |||
| FQ215499.1 | - | 1 | 28 | |||
| FQ218937.1 | - | 1 | 24 | |||
| FQ219058.1 | - | 1 | 24 | |||
| FQ222789.1 | - | 1 | 28 | |||

Gltscr2
Genomic Map